This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01279080_m1
See other RHOG GE Assays ›
Gene Symbol:
Rhog
Gene Name:
ras homolog gene family, member G
Gene Aliases:
2810426G09Rik, Arhg, Sid10750
Chromosome Location:
Chr.7: 102239123 - 102250118 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01279080_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rhog
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019566.3 NP_062512.1 1 - 2 79 108
GenBank mRNA BQ942269.1 - 1 - 2 79 108

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_019566.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rhog

Panther Classification:

Molecular Function -

small GTPase

Gene Ontology Categories:

Function(s) Process(es)

small GTPase mediated signal transduction
Rho protein signal transduction
metabolic process
Rac protein signal transduction
actin cytoskeleton organization
positive regulation of transcription, DNA-templated
cell chemotaxis
positive regulation of establishment of protein localization to plasma membrane
activation of GTPase activity
regulation of ruffle assembly
nucleotide binding
GTPase activity
protein binding
GTP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700032620.mm.1 NM_019566.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700032620.mm.1 NM_019566.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700032620.mm.1 TC0700003857.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700032622.mm.1 NM_019566.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700032622.mm.1 NM_019566.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700032622.mm.1 TC0700003857.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0