This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01278760_m1
See other CHAF1A GE Assays ›
Gene Symbol:
Chaf1a
Gene Name:
chromatin assembly factor 1, subunit A (p150)
Gene Aliases:
AL023013, AL024058, CAF-1, Cac1p
Chromosome Location:
Chr.17: 56040416 - 56068026 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01278760_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Chaf1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013733.3 NP_038761.1 3 - 4 998 108
GenBank mRNA AJ132771.2 - 3 - 4 902 108
AK034839.1 - 3 - 4 999 108
AK132551.1 - 822 108
AK135763.1 - 3 - 4 1011 108
BC053740.1 - 3 - 4 969 6504838 108

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700000997.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Chaf1a

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

DNA replication
DNA repair
nucleosome assembly
DNA replication-dependent nucleosome assembly
cellular response to DNA damage stimulus
cell cycle
chromatin assembly
protein binding
identical protein binding
chromo shadow domain binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700009519.mm.1 NM_013733.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009519.mm.1 NM_013733.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700009519.mm.1 TC1700000997.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700009518.mm.1 NM_013733.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009518.mm.1 NM_013733.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700009518.mm.1 TC1700000997.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0