This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01277314_m1
See other PAX6OS1 GE Assays ›
Gene Symbol:
Pax6os1
Gene Name:
paired box 6 opposite strand 1
Gene Aliases:
4831410G09Rik, Pax6os
Chromosome Location:
Chr.2: 105536080 - 105670330 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01277314_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pax6os1
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_002867.2
1 - 2 93 76
GenBank mRNA AK029183.1 - LIT3400
Pax6OS
1 - 2 94 76
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB613704.1 - 1 - 2 94 Chr. 2 - 105536939 - 105670330 [-] on Build GRCm38 76

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream Pax6 upstream antisense RNA Paupar Sense Chr. 2 - 105657862 - 105661343 on NCBI Build GRCm38
Downstream elongation protein 4 homolog (S. cerevisiae) Elp4 A330107A17Rik,
Paxneb
29.8kb Sense Chr. 2 - 105700109 - 105904500 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200004265.mm.1

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200035885.mm.1 NR_002867.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200035885.mm.1 NR_002867.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200035885.mm.1 TC0200004265.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200035889.mm.1 NR_002867.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200035889.mm.1 NR_002867.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200035889.mm.1 TC0200004265.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200035888.mm.1 NR_002867.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200035888.mm.1 NR_002867.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200035888.mm.1 TC0200004265.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0