This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01276490_m1
See other SLC24A3 GE Assays ›
Gene Symbol:
Slc24a3
Gene Name:
solute carrier family 24 (sodium/potassium/calcium exchanger), member 3
Gene Aliases:
NCKX3
Chromosome Location:
Chr.2: 145167712 - 145642166 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01276490_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc24a3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053195.2 NP_444425.1 2 - 3 173 80
GenBank mRNA AF314821.1 - 2 - 3 213 80
BC017615.1 - 2 - 3 173 4914291 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002157.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc24a3

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
cation transport
potassium ion transport
sodium ion transport
calcium ion transport
cellular calcium ion homeostasis
sodium ion transmembrane transport
transmembrane transport
calcium ion transmembrane transport
potassium ion transmembrane transport
calcium, potassium:sodium antiporter activity
symporter activity
antiporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0