This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01275854_g1
See other GPD2 GE Assays ›
Gene Symbol:
Gpd2
Gene Name:
glycerol phosphate dehydrogenase 2, mitochondrial
Gene Aliases:
AA408484, AI448216, AU021455, AW494132, GPDH, Gdm1, Gpd-m, Gpdh-m, TISP38
Chromosome Location:
Chr.2: 57237678 - 57370719 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01275854_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpd2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001145820.1 NP_001139292.1 11 - 12 1796 68
NM_010274.3 NP_034404.3 11 - 12 1671 68
GenBank mRNA AK034353.1 - 11 - 12 1580 68
AK144716.1 - 11 - 12 1797 68
AK151189.1 - 11 - 12 1584 68
AK167152.1 - 11 - 12 1609 68
BC021359.1 - 11 - 12 1765 5012142 68
U60987.1 - 11 - 12 1607 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000868.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpd2

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

glycerol-3-phosphate metabolic process
gluconeogenesis
NADH metabolic process
multicellular organism growth
camera-type eye development
oxidation-reduction process
glycerol-3-phosphate dehydrogenase [NAD+] activity
glycerol-3-phosphate dehydrogenase activity
calcium ion binding
oxidoreductase activity
metal ion binding
sn-glycerol-3-phosphate:ubiquinone oxidoreductase activity
sn-glycerol-3-phosphate:ubiquinone-8 oxidoreductase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200007022.mm.1 NM_001145820.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007022.mm.1 NM_001145820.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007022.mm.1 NM_010274.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007022.mm.1 NM_010274.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200007022.mm.1 TC0200000868.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0