This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01275725_m1
See other ACADSB GE Assays ›
Gene Symbol:
Acadsb
Gene Name:
acyl-Coenzyme A dehydrogenase, short/branched chain
Gene Aliases:
1300003O09Rik, 2-MEBCAD, BB066609, SBCAD
Chromosome Location:
Chr.7: 131410601 - 131446211 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01275725_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Acadsb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025826.4 NP_080102.1 7 - 8 1040 118
GenBank mRNA AK004889.1 - 7 - 8 941 118
AK076535.1 - 7 - 8 941 118
AK131628.1 - 7 - 8 987 118
AK152325.1 - 7 - 8 929 118
AK163322.1 - 3 - 4 521 118
BC011235.1 - 7 - 8 922 118
BC035231.1 - 7 - 8 906 118
BC044826.1 - 7 - 8 973 118
BC054428.1 - 7 - 8 982 5039627 118

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700001922.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Acadsb

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid metabolic process
acyl-CoA metabolic process
metabolic process
oxidation-reduction process
acyl-CoA dehydrogenase activity
electron carrier activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-CH group of donors
short-branched-chain-acyl-CoA dehydrogenase activity
flavin adenine dinucleotide binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700016877.mm.1 NM_025826.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700016877.mm.1 NM_025826.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700016877.mm.1 TC0700001922.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0