This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01275342_m1
See other GDPD3 GE Assays ›
Gene Symbol:
Gdpd3
Gene Name:
glycerophosphodiester phosphodiesterase domain containing 3
Gene Aliases:
1110015E22Rik
Chromosome Location:
Chr.7: 126766414 - 126775645 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01275342_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gdpd3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024228.2 NP_077190.2 9 - 10 875 110
GenBank mRNA AK003726.1 - 9 - 10 877 110
AW045753.1 - 1 - 2 110
BC002172.1 - 7 - 8 794 110

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700001815.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gdpd3

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

glycerol metabolic process
lipid metabolic process
nuclease activity
phosphoric diester hydrolase activity
glycerophosphodiester phosphodiesterase activity
hydrolase activity
phosphoric ester hydrolase activity
T/G mismatch-specific endonuclease activity
retroviral 3' processing activity
metal ion binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426875 NM_024228.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426875 AK003726.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426875 BC002172.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426875 NM_024228.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426875 AK003726.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426875 BC002172.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 528469 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 167711 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 195101 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 873462 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 438491 AK009290.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 963800 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 96202 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150950 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426875 NM_024228.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426875 AK003726.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426875 BC002172.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426875 NM_024228.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426875 AK003726.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426875 BC002172.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426875 NM_024228.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426875 AK003726.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426875 BC002172.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426875 NM_024228.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426875 AK003726.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426875 BC002172.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502116 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502081 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502120 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502091 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502103 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502095 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502094 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502099 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502086 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502088 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502128 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502082 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502096 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502098 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502102 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502125 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502129 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502092 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502123 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502093 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502122 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502097 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502087 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502084 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502100 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502118 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01630259 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01630266 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P150950 - -
Whole Mouse Genome G4122A A_51_P426875 NM_024228.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P426875 AK003726.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P426875 BC002172.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P426875 NM_024228.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P426875 AK003726.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P426875 BC002172.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134804 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P150950 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06273527 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06273517 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06273535 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06273519 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06273526 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502118 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502188 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502107 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502137 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502049 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502102 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502087 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502109 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502038 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502030 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502175 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502100 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502131 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502120 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502197 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502088 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502042 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502203 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502044 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502084 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502166 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502124 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502037 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502035 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502181 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502204 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502199 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502079 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502050 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502190 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502162 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502052 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502186 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502101 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502164 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502054 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502095 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502183 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502126 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502093 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502117 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502086 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502146 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502132 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502134 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502135 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502170 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502139 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502129 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502191 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502179 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502091 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502110 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502113 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502122 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502130 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502169 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502148 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502115 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502040 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502195 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502114 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502099 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502039 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502082 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502155 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502032 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502185 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502150 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502153 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502174 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502173 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502034 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502206 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502098 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502201 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502142 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502171 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502096 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502161 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502128 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502094 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502111 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06273504 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502111 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06072354 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06653404 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06644101 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06273489 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502116 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06388981 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502086 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502107 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502122 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502093 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502101 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06147818 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502110 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502082 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502102 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06273490 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06273513 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06521864 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502113 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502117 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502091 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502118 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502115 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06644098 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502129 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502123 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502094 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502109 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06273537 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06629780 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06273499 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502098 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10557480 - -
Affymetrix Mouse Gene 1.0 ST Array 10557478 - -
Affymetrix Mouse Gene 1.0 ST Array 10557471 - -
Affymetrix Mouse Gene 1.0 ST Array 10557475 - -
Affymetrix Mouse Gene 1.0 ST Array 10557477 - -
Affymetrix Mouse Gene 1.0 ST Array 10557479 - -
Affymetrix Mouse Gene 1.0 ST Array 10557472 - -
Affymetrix Mouse Gene 1.0 ST Array 10557476 - -
Affymetrix Mouse Gene 1.0 ST Array 10557473 - -
Affymetrix Mouse Gene 1.0 ST Array 10557474 - -
Affymetrix HT MG-430 PM Array Plate 1449526_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10557470 - -
Mouse Genome 430 2.0 Array 1449526_PM_a_at - -
Mouse Genome 430 2.0 Array 1449526_a_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 68616_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 68616_at - -
Affymetrix Mouse Clariom S Array PSR0700015493.mm.1 NM_024228.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700015493.mm.1 NM_024228.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700015493.mm.1 TC0700001815.mm.1 MTA TC
Mouse Expression Set 430 1449526_PM_a_at - -
Mouse Expression Set 430 1449526_a_at - -
Murine Genome U74v2 Set 104217_PM_at - -
Murine Genome U74v2 Set 104217_at - -
Mouse Genome 430A 2.0 Array 1449526_a_at - -
Mouse Genome 430A 2.0 Array 1449526_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1449526_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1449526_a_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 6110292 - -
Illumina mouse-6 v1.0 expression beadchip 6110292 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006110292 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2685882 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2685882 - -
mouse-6 v1.1 (Illumina) scl32054.9.1_17-S - -
Illumina Sentrix mouse-6.v1 bead array 6110292 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl32054.9.1_17-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6110292 - -
MouseRef-8 v2 Expression BeadChip ILMN_2893879 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2893879 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0