This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01275005_m1
See other ZFP143 GE Assays ›
Gene Symbol:
Zfp143
Gene Name:
zinc finger protein 143
Gene Aliases:
AA959806, AU015869, D7Ertd805e, KRAB14, SBF, Staf, Zfp79, Zfp80-rs1, Znf143, pHZ-1
Chromosome Location:
Chr.7: 110061670 - 110095396 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01275005_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zfp143
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009281.3 NP_033307.2 1 - 2 69 101
GenBank mRNA AK029078.1 - 1 - 2 61 101
BC037658.1 - 1 - 2 60 5356405 101
CN709211.1 - 1 - 2 69 101

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK029078.1, BC037658.1, NM_009281.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Zfp143

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of smooth muscle cell proliferation
nucleic acid binding
DNA binding
chromatin binding
protein binding
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700013443.mm.1 NM_009281.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700013443.mm.1 NM_009281.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700013443.mm.1 TC0700001623.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700013444.mm.1 NM_009281.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700013444.mm.1 NM_009281.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700013444.mm.1 TC0700001623.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 565000 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0