This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01275003_m1
See other ZFP143 GE Assays ›
Gene Symbol:
Zfp143
Gene Name:
zinc finger protein 143
Gene Aliases:
AA959806, AU015869, D7Ertd805e, KRAB14, SBF, Staf, Zfp79, Zfp80-rs1, Znf143, pHZ-1
Chromosome Location:
Chr.7: 110061670 - 110095396 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01275003_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zfp143
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009281.3 NP_033307.2 7 - 8 719 89
GenBank mRNA AF011758.2 - 668 89
AK028056.1 - 7 - 8 712 89
AK029078.1 - 7 - 8 711 89
AK029926.1 - 7 - 8 984 89
BC037658.1 - 6 - 7 629 5356405 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700001623.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Zfp143

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of smooth muscle cell proliferation
nucleic acid binding
DNA binding
chromatin binding
protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700013451.mm.1 NM_009281.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700013451.mm.1 NM_009281.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700013451.mm.1 TC0700001623.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0