This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01273407_m1
See other DNER GE Assays ›
Gene Symbol:
Dner
Gene Name:
delta/notch-like EGF-related receptor
Gene Aliases:
A930026D19Rik, BET, Bret
Chromosome Location:
Chr.1: 84369839 - 84696221 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01273407_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dner
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_152915.1 NP_690879.1 2 - 3 1079 122
GenBank mRNA AB067650.1 - 2 - 3 728 122
AF370126.1 - 2 - 3 735 122
AK030642.1 - 2 - 3 878 122
AK044597.1 - 2 - 3 842 122
AY032924.1 - 2 - 3 735 122
BC034634.1 - 2 - 3 1079 5365190 122
BC116231.1 - 1 - 2 118 122
BC128476.1 - 1 - 2 118 122

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100002696.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dner

Gene Ontology Categories:

Function(s) Process(es)

Notch signaling pathway
Notch receptor processing
multicellular organism development
glial cell differentiation
cell differentiation
skeletal muscle fiber development
Notch binding
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100021859.mm.1 NM_152915.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100021859.mm.1 NM_152915.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100021859.mm.1 TC0100002696.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0