This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01271512_m1
See other FAR1 GE Assays ›
Gene Symbol:
Far1
Gene Name:
fatty acyl CoA reductase 1
Gene Aliases:
2600011M19Rik, 2900034E22Rik, 3732409C05Rik, AI850429, Mlstd2
Chromosome Location:
Chr.7: 113513834 - 113571506 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01271512_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Far1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027379.3 NP_081655.2 9 - 10 1114 82
GenBank mRNA AK005531.1 - 9 - 10 1088 82
AK014405.1 - 9 - 10 1087 82
AK014486.1 - 9 - 10 1087 82
AK030067.1 - 9 - 10 1045 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700001666.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Far1

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
ether lipid biosynthetic process
wax biosynthetic process
long-chain fatty-acyl-CoA metabolic process
glycerophospholipid biosynthetic process
oxidation-reduction process
oxidoreductase activity
long-chain-fatty-acyl-CoA reductase activity
fatty-acyl-CoA reductase (alcohol-forming) activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700013854.mm.1 NM_027379.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700013854.mm.1 TC0700001666.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0