This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01270960_m1
See other MYEF2 GE Assays ›
Gene Symbol:
Myef2
Gene Name:
myelin basic protein expression factor 2, repressor
Gene Aliases:
9430071B01, mKIAA1341
Chromosome Location:
Chr.2: 125082209 - 125123661 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01270960_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Myef2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001162417.1 NP_001155889.1 18 - 19 1957 111
GenBank mRNA AK048404.1 - 18 - 19 1924 111

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200004521.mm.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein RNA processing factor RNA splicing factor nucleic acid metabolism protein

Gene Ontology Categories:

Process(es)

poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200038403.mm.1 NM_001162417.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200038403.mm.1 NM_001162417.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200038403.mm.1 TC0200004521.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200038404.mm.1 NM_001162417.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200038404.mm.1 NM_001162417.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200038404.mm.1 TC0200004521.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0