This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01270222_m1
See other FAM49A GE Assays ›
Gene Symbol:
Fam49a
Gene Name:
family with sequence similarity 49, member A
Gene Aliases:
2410157M17Rik, 9630047E15, D12Ertd553e
Chromosome Location:
Chr.12: 12262129 - 12379058 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01270222_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fam49a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001146119.1 NP_001139591.1 5 - 6 552 82
NM_029758.4 NP_084034.2 6 - 7 625 82
GenBank mRNA AK047142.1 - 5 - 6 515 82
BC049664.1 - 6 - 7 587 6532813 82
BC052465.1 - 5 - 6 533 6834768 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200000098.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fam49a

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200000966.mm.1 NM_001146119.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200000966.mm.1 NM_001146119.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200000966.mm.1 NM_029758.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200000966.mm.1 NM_029758.4 Refseq
Affymetrix Mouse Clariom S Array PSR1200000966.mm.1 TC1200000098.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0