This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01269506_g1
See other SEC24C GE Assays ›
Gene Symbol:
Sec24c
Gene Name:
Sec24 related gene family, member C (S. cerevisiae)
Gene Aliases:
2610204K03Rik, mKIAA0079
Chromosome Location:
Chr.14: 20674285 - 20694852 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01269506_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sec24c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001168273.1 NP_001161745.1 10 - 11 1233 75
NM_172596.2 NP_766184.2 9 - 10 1461 75
GenBank mRNA AK036519.1 - 9 - 10 1461 75
AK122199.1 - 10 - 11 1211 75
AK132090.1 - 4 - 5 488 75
BC040370.1 - 9 - 10 1439 4239204 75

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sec24c

Panther Classification:

Molecular Function -

vesicle coat protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0