This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01268813_m1
See other DTWD1 GE Assays ›
Gene Symbol:
Dtwd1
Gene Name:
DTW domain containing 1
Gene Aliases:
1810033A06Rik
Chromosome Location:
Chr.2: 126152113 - 126165277 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01268813_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dtwd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026981.2 NP_081257.1 1 - 2 75 90
GenBank mRNA AK149746.1 - 1 - 2 76 90
BY240339.1 - 1 - 2 78 90

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_026981.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dtwd1

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200014739.mm.1 NM_026981.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200014739.mm.1 NM_026981.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200014739.mm.1 TC0200001895.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200014744.mm.1 NM_026981.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200014744.mm.1 NM_026981.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200014744.mm.1 TC0200001895.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0