This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01268344_m1
See other ARHGAP35 GE Assays ›
Gene Symbol:
Arhgap35
Gene Name:
Rho GTPase activating protein 35
Gene Aliases:
6430596G11Rik, AI841135, Grlf1, mKIAA1722, p190A, p190RhoGAP
Chromosome Location:
Chr.7: 16494473 - 16614993 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01268344_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap35
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172739.4 NP_766327.3 3 - 4 4536 88
GenBank mRNA AK136766.1 - 2 - 3 1210 88
AK147326.1 - 3 - 4 4536 88
AK147688.1 - 3 - 4 4536 88
AK173242.1 - 2 - 3 3338 88
BC035541.1 - 2 - 3 599 88
BC139459.1 - 4 - 5 4314 8862481 88
BC139461.1 - 4 - 5 4173 8862483 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002430.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arhgap35

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
neural tube closure
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
signal transduction
integrin-mediated signaling pathway
small GTPase mediated signal transduction
regulation of cell shape
positive regulation of neuron projection development
forebrain development
negative regulation of Rho protein signal transduction
camera-type eye development
negative regulation of vascular permeability
positive regulation of GTPase activity
negative regulation of transcription, DNA-templated
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
GTPase activator activity
GTP binding
protein complex binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700021417.mm.1 NM_172739.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700021417.mm.1 NM_172739.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700021417.mm.1 TC0700002430.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0