This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01265992_g1
See other PNN GE Assays ›
Gene Symbol:
Pnn
Gene Name:
pinin
Gene Aliases:
AU045199, D12Ertd512e
Chromosome Location:
Chr.12: 59066919 - 59074017 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01265992_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pnn
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008891.2 NP_032917.2 2 - 3 273 88
GenBank mRNA AK009055.1 - 2 - 3 228 88
AK028219.1 - 2 - 3 214 88
AK030978.1 - 2 - 3 202 88
AK050669.1 - 2 - 3 219 88
AK051640.1 - 2 - 3 271 88
AK085295.1 - 2 - 3 89
AK157587.1 - 2 - 3 217 88
AK160616.1 - 2 - 3 208 88
AK161109.1 - 2 - 3 213 88
BC132571.1 - 2 - 3 207 40130848 88
BC138014.1 - 2 - 3 207 8861033 88
BY775218.1 - 2 - 3 274 88
Y08701.1 - 2 - 3 207 88

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pnn

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
RNA splicing
single organismal cell-cell adhesion
DNA binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0