Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100020211.mm.1 |
NM_001122738.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020211.mm.1 |
NM_001122738.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020211.mm.1 |
NM_177645.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020211.mm.1 |
NM_177645.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020211.mm.1 |
TC0100002488.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100020225.mm.1 |
NM_001122738.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020225.mm.1 |
NM_001122738.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020225.mm.1 |
NM_177645.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020225.mm.1 |
NM_177645.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020225.mm.1 |
TC0100002488.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100020209.mm.1 |
NM_177645.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020209.mm.1 |
NM_177645.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020209.mm.1 |
TC0100002488.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100020219.mm.1 |
NM_001122738.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020219.mm.1 |
NM_001122738.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020219.mm.1 |
NM_177645.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020219.mm.1 |
NM_177645.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020219.mm.1 |
TC0100002488.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100020218.mm.1 |
NM_001122738.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020218.mm.1 |
NM_001122738.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020218.mm.1 |
NM_177645.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020218.mm.1 |
NM_177645.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020218.mm.1 |
TC0100002488.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100020227.mm.1 |
NM_001122738.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020227.mm.1 |
NM_001122738.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020227.mm.1 |
NM_177645.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020227.mm.1 |
NM_177645.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020227.mm.1 |
TC0100002488.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100020223.mm.1 |
NM_001122738.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020223.mm.1 |
NM_001122738.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020223.mm.1 |
NM_177645.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020223.mm.1 |
NM_177645.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020223.mm.1 |
TC0100002488.mm.1 |
MTA TC |