This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01262296_m1
See other CAMK2A GE Assays ›
Gene Symbol:
Camk2a
Gene Name:
calcium/calmodulin-dependent protein kinase II alpha
Gene Aliases:
CaMKII, R74975, mKIAA0968
Chromosome Location:
Chr.18: 60925580 - 60988152 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01262296_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Camk2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009792.3 NP_033922.1 2 - 3 439 82
GenBank mRNA X87142.1 - 2 - 3 439 82

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Protein Phosphatase Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Camk2a

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
response to ischemia
protein phosphorylation
calcium ion transport
positive regulation of cardiac muscle cell apoptotic process
phosphorylation
peptidyl-serine phosphorylation
ionotropic glutamate receptor signaling pathway
angiotensin-activated signaling pathway
protein autophosphorylation
regulation of neurotransmitter secretion
regulation of neuronal synaptic plasticity
positive regulation of NF-kappaB transcription factor activity
positive regulation of calcium ion transport
regulation of mitochondrial membrane permeability involved in apoptotic process
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
protein binding
calmodulin binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
GTPase activating protein binding
glutamate receptor binding
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0