This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01260260_m1
See other KCNAB2 GE Assays ›
Gene Symbol:
Kcnab2
Gene Name:
potassium voltage-gated channel, shaker-related subfamily, beta member 2
Gene Aliases:
F5, I2rf5, Kcnb3, kv-beta-2
Chromosome Location:
Chr.4: 152390740 - 152477549 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01260260_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Kcnab2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252654.1 NP_001239583.1 4 - 5 360 82
NM_001252655.1 NP_001239584.1 4 - 5 389 82
NM_001252656.1 NP_001239585.1 5 - 6 431 82
NM_010598.3 NP_034728.2 5 - 6 402 82
GenBank mRNA AK143166.1 - 4 - 5 337 82
AK152613.1 - 5 - 6 354 82
AK157053.1 - 4 - 5 353 82
BC039178.1 - 5 - 6 332 4503373 82
L48983.1 - 5 - 6 394 82
U31908.1 - 5 - 6 339 82
U65592.1 - 4 - 5 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400004097.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Kcnab2

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
transport
ion transport
potassium ion transport
regulation of ion transmembrane transport
neuromuscular process
protein heterooligomerization
oxidation-reduction process
NADPH oxidation
regulation of potassium ion transmembrane transport
regulation of protein localization to cell surface
C-3 sterol dehydrogenase (C-4 sterol decarboxylase) activity
aldo-keto reductase (NADP) activity
isocitrate dehydrogenase activity
mevaldate reductase activity
voltage-gated ion channel activity
voltage-gated potassium channel activity
protein binding
gluconate dehydrogenase activity
potassium channel regulator activity
steroid dehydrogenase activity
oxidoreductase activity
epoxide dehydrogenase activity
5-exo-hydroxycamphor dehydrogenase activity
2-hydroxytetrahydrofuran dehydrogenase activity
acetoin dehydrogenase activity
phenylcoumaran benzylic ether reductase activity
D-xylose:NADP reductase activity
L-arabinose:NADP reductase activity
D-arabinitol dehydrogenase, D-ribulose forming (NADP+) activity
steroid dehydrogenase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
steroid dehydrogenase activity, acting on the CH-CH group of donors
(R)-(-)-1,2,3,4-tetrahydronaphthol dehydrogenase activity
3-hydroxymenthone dehydrogenase activity
very long-chain-3-hydroxyacyl-CoA dehydrogenase activity
dihydrotestosterone 17-beta-dehydrogenase activity
(R)-2-hydroxyisocaproate dehydrogenase activity
L-arabinose 1-dehydrogenase (NADP+) activity
L-xylulose reductase (NAD+) activity
ion channel binding
3-ketoglucose-reductase activity
(R)-2-hydroxyglutarate dehydrogenase activity
D-arabinitol dehydrogenase, D-xylulose forming (NADP+) activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400034191.mm.1 NM_001252655.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400034191.mm.1 NM_001252656.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400034191.mm.1 TC0400004097.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400034190.mm.1 NM_001252655.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400034190.mm.1 NM_001252656.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400034190.mm.1 TC0400004097.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400034172.mm.1 NM_001252654.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400034172.mm.1 NM_001252655.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400034172.mm.1 NM_001252656.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400034172.mm.1 NM_010598.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400034172.mm.1 NM_010598.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400034172.mm.1 TC0400004097.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0