This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01258784_m1
See other ABLIM3 GE Assays ›
Gene Symbol:
Ablim3
Gene Name:
actin binding LIM protein family, member 3
Gene Aliases:
AW990719, D930036B08Rik
Chromosome Location:
Chr.18: 61799392 - 61911857 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01258784_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ablim3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164491.1 NP_001157963.1 11 - 12 1309 81
NM_198649.3 NP_941051.2 10 - 11 1628 81
GenBank mRNA AK086538.1 - 10 - 11 1408 81
AK173040.1 - 10 - 11 1186 81
BC060275.1 - 11 - 12 1280 6822626 81
BC094229.1 - 10 - 11 1602 30534236 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800001431.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ablim3

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
cytoskeleton organization
lamellipodium assembly
actin cytoskeleton organization
cilium assembly
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein targeting to mitochondrion
molecular_function
actin binding
zinc ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800010535.mm.1 NM_001164491.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800010535.mm.1 NM_001164491.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800010535.mm.1 NM_198649.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800010535.mm.1 NM_198649.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800010535.mm.1 TC1800001431.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0