This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01258018_m1
See other 4930552P12RIK GE Assays ›
Gene Symbol:
4930552P12Rik
Gene Name:
RIKEN cDNA 4930552P12 gene
Gene Aliases:
-
Chromosome Location:
Chr.19: 55640589 - 55702204 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01258018_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930552P12Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045318.1 2 - 3 360 64
GenBank mRNA AK019762.1 - 2 - 3 361 64
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY715703.1 - 2 - 3 361 Chr. 19 - 55642420 - 55702204 [-] on Build GRCm38 64

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream vesicle transport through interaction with t-SNAREs 1A Vti1a 1110014F16Rik,
1110018K19Rik,
4921537J05Rik,
MVti1,
MVti1a,
Vti1,
Vti1-rp2
-16.0kb Anti-sense Chr. 19 - 55316057 - 55627461 on NCBI Build GRCm38
Downstream transcription factor 7 like 2,
T cell specific,
HMG box
Tcf7l2 TCF4B,
TCF4E,
Tcf-4,
Tcf4,
mTcf-4B,
mTcf-4E
43.2kb Anti-sense Chr. 19 - 55741810 - 55933661 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900001699.mm.1
This assay targets the same location as RTA transcript cluster(s) TC0100003843.rn

Set Membership:

Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900014914.mm.1 NR_045318.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900014914.mm.1 NR_045318.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900014914.mm.1 TC1900001699.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900014915.mm.1 NR_045318.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900014915.mm.1 NR_045318.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900014915.mm.1 TC1900001699.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900014916.mm.1 NR_045318.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900014916.mm.1 NR_045318.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900014916.mm.1 TC1900001699.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0