This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01257079_m1
See other IDH1 GE Assays ›
Gene Symbol:
Idh1
Gene Name:
isocitrate dehydrogenase 1 (NADP+), soluble
Gene Aliases:
AI314845, AI788952, E030024J03Rik, Id-1, Idh-1, Idpc
Chromosome Location:
Chr.1: 65158616 - 65186479 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01257079_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Idh1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001111320.1 NP_001104790.1 5 - 6 891 101
NM_010497.3 NP_034627.3 5 - 6 869 101
GenBank mRNA AF020039.1 - 4 - 5 719 101
AK087063.1 - 5 - 6 771 101
AK149019.1 - 5 - 6 800 101
AK151212.1 - 5 - 6 794 101
AK159173.1 - 5 - 6 800 101
AK160896.1 - 5 - 6 801 101
AK167158.1 - 5 - 6 841 101
AK167583.1 - 5 - 6 805 101
AK168832.1 - 5 - 6 843 101
BC088986.1 - 5 - 6 841 6808731 101
DQ403075.1 - 4 - 5 619 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100002481.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Idh1

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

glyoxylate cycle
tricarboxylic acid cycle
isocitrate metabolic process
2-oxoglutarate metabolic process
glutathione metabolic process
response to oxidative stress
response to organic cyclic compound
response to steroid hormone
oxidation-reduction process
regulation of phospholipid catabolic process
regulation of phospholipid biosynthetic process
magnesium ion binding
isocitrate dehydrogenase (NADP+) activity
receptor binding
oxidoreductase activity
oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
protein homodimerization activity
metal ion binding
NADP binding
NAD binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0