This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01256455_m1
See other ACADL GE Assays ›
Gene Symbol:
Acadl
Gene Name:
acyl-Coenzyme A dehydrogenase, long-chain
Gene Aliases:
AA960361, AU018452, C79855, LCAD
Chromosome Location:
Chr.1: 66830839 - 66863309 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01256455_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Acadl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007381.4 NP_031407.2 1 - 2 136 136
GenBank mRNA AK151845.1 - 1 - 2 105 136
AK167537.1 - 1 - 2 105 136

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_007381.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Acadl

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

temperature homeostasis
lipid metabolic process
fatty acid metabolic process
metabolic process
carnitine metabolic process, CoA-linked
fatty acid beta-oxidation using acyl-CoA dehydrogenase
carnitine catabolic process
long-chain fatty acid catabolic process
cellular lipid catabolic process
negative regulation of fatty acid biosynthetic process
negative regulation of fatty acid oxidation
protein homotetramerization
oxidation-reduction process
regulation of cholesterol metabolic process
fatty-acyl-CoA binding
acyl-CoA dehydrogenase activity
long-chain-acyl-CoA dehydrogenase activity
palmitoyl-CoA oxidase activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-CH group of donors
flavin adenine dinucleotide binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100020248.mm.1 NM_007381.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100020248.mm.1 TC0100002495.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100020246.mm.1 NM_007381.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100020246.mm.1 TC0100002495.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100020251.mm.1 NM_007381.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100020251.mm.1 TC0100002495.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0