This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01253067_m1
See other 1700016P03RIK GE Assays ›
Gene Symbol:
1700016P03Rik
Gene Name:
RIKEN cDNA 1700016P03 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01253067_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1700016P03Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK006051.1 - 219 72
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY706166.1 - 2 - 3 219 Chr. 11 - 75172560 - 75177623 [+] on Build GRCm38 72

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream candidate tumor suppressor in ovarian cancer 2 Ovca2 9030414M07Rik Anti-sense Chr. 11 - 75175943 - 75178808 on NCBI Build GRCm38
Downstream DPH1 homolog (S. cerevisiae) Dph1 2310011M22Rik,
4930488F09Rik,
AW551873,
Dph2l1,
Ovca1
20.0 Anti-sense Chr. 11 - 75177643 - 75190483 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

MicroArray Probes:

Agilent ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2185526 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 382554 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164872 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135833 - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 342753 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 107000040 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0