This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01250866_m1
See other DVL2 GE Assays ›
Gene Symbol:
Dvl2
Gene Name:
dishevelled 2, dsh homolog (Drosophila)
Gene Aliases:
-
Chromosome Location:
Chr.11: 70000626 - 70010109 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01250866_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dvl2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007888.3 NP_031914.3 9 - 10 1241 72
GenBank mRNA AK144001.1 - 9 - 10 1272 72
AK146822.1 - 9 - 10 1241 72
AK159895.1 - 9 - 10 1074 72
AK168376.1 - 9 - 10 1149 72
AK219360.1 - 3 - 4 296 72
BC053050.1 - 9 - 10 1155 6402000 72
BC092396.1 - 1248 30363483 72

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dvl2

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

neural tube closure
positive regulation of protein phosphorylation
heart morphogenesis
outflow tract morphogenesis
transcription from RNA polymerase II promoter
multicellular organism development
segment specification
heart development
Wnt signaling pathway
convergent extension involved in neural plate elongation
cellular protein localization
segmentation
intracellular signal transduction
non-canonical Wnt signaling pathway
positive regulation of JUN kinase activity
canonical Wnt signaling pathway involved in regulation of cell proliferation
positive regulation of transcription, DNA-templated
positive regulation of sequence-specific DNA binding transcription factor activity
convergent extension involved in organogenesis
canonical Wnt signaling pathway
Wnt signaling pathway, planar cell polarity pathway
positive regulation of protein tyrosine kinase activity
cochlea morphogenesis
planar cell polarity pathway involved in neural tube closure
positive regulation of canonical Wnt signaling pathway
frizzled binding
protein binding
protein domain specific binding
identical protein binding
protein self-association

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100008111.mm.1 NM_007888.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100008111.mm.1 NM_007888.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100008111.mm.1 TC1100000936.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0