This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01249522_m1
See other NCOA3 GE Assays ›
Gene Symbol:
Ncoa3
Gene Name:
nuclear receptor coactivator 3
Gene Aliases:
2010305B15Rik, AW321064, Actr, Aib1, KAT13B, Rac3, Src3, Tram-1, Tram1, bHLHe42, p/Cip, pCip
Chromosome Location:
Chr.2: 165992637 - 166073242 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01249522_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ncoa3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008679.3 NP_032705.2 10 - 11 1320 74
GenBank mRNA AK137507.1 - 10 - 11 1306 74
AK142306.1 - 10 - 11 1225 74
AK156345.1 - 10 - 11 1298 74
BC086482.1 - 10 - 11 1290 30104508 74
BC093478.1 - 7 - 8 951 74
BC141177.1 - 9 - 10 1151 9055789 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002558.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ncoa3

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
signal transduction
positive regulation of gene expression
histone acetylation
positive regulation of cell growth
cellular response to hormone stimulus
positive regulation of intracellular steroid hormone receptor signaling pathway
positive regulation of intracellular estrogen receptor signaling pathway
multicellular organism growth
negative regulation of apoptotic process
positive regulation of keratinocyte differentiation
positive regulation of transcription from RNA polymerase II promoter
developmental growth
positive regulation of sequence-specific DNA binding transcription factor activity
vagina development
labyrinthine layer morphogenesis
mammary gland branching involved in thelarche
cellular response to estradiol stimulus
regulation of RNA biosynthetic process
RNA polymerase II regulatory region DNA binding
chromatin binding
transcription coactivator activity
histone acetyltransferase activity
signal transducer activity
transferase activity
transferase activity, transferring acyl groups
ligand-dependent nuclear receptor binding
estrogen receptor binding
ligand-dependent nuclear receptor transcription coactivator activity
protein complex binding
nuclear hormone receptor binding
peroxisome proliferator activated receptor binding
protein dimerization activity
protein N-terminus binding
androgen receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200021000.mm.1 NM_008679.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200021000.mm.1 NM_008679.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200021000.mm.1 TC0200002558.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0