This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01249475_m1
See other C330019F10RIK GE Assays ›
Gene Symbol:
C330019F10Rik
Gene Name:
RIKEN cDNA C330019F10 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01249475_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C330019F10Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK049282.1 - 2 - 3 90 91
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB400623.2 - 2 - 3 127 Chr. 11 - 107511401 - 107527223 [-] on Build GRCm38 91
GenBankmRNA BB651899.1 - 2 - 3 90 Chr. 11 - 107511400 - 107527220 [-] on Build GRCm38 91

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RIKEN cDNA A830035A12 gene A830035A12Rik Sense Chr. 11 - 107526506 - 107534143 on NCBI Build GRCm38
Downstream helicase with zinc finger domain Helz 3110078M01Rik,
9430093I07Rik,
9630002H22Rik,
AI851979,
DRHC
20.4kb Anti-sense Chr. 11 - 107546958 - 107693826 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0