This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01249439_m1
See other RPTOROS GE Assays ›
Gene Symbol:
Rptoros
Gene Name:
regulatory associated protein of MTOR, complex 1, opposite strand
Gene Aliases:
A430071A18Rik
Chromosome Location:
Chr.11: 119820811 - 119834365 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01249439_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rptoros
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045312.1 1 - 2 73 74
GenBank mRNA AK040159.1 - 1 - 2 65 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100004189.mm.1

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100039791.mm.1 NR_045312.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100039791.mm.1 NR_045312.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100039791.mm.1 TC1100004189.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100039792.mm.1 NR_045312.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100039792.mm.1 NR_045312.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100039792.mm.1 NR_045313.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100039792.mm.1 NR_045313.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100039792.mm.1 TC1100004189.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0