This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01247962_m1
See other 1110018N20RIK GE Assays ›
Gene Symbol:
1110018N20Rik
Gene Name:
RIKEN cDNA 1110018N20 gene
Gene Aliases:
1190017L20Rik
Chromosome Location:
Chr.2: 167188729 - 167202996 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01247962_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1110018N20Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK004537.1 - 1 - 2 301 126
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY704762.1 - 1 - 2 298 Chr. 2 - 167189109 - 167192494 [+] on Build GRCm38 126

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream potassium voltage gated channel,
Shab-related subfamily,
member 1
Kcnb1 Kcr1-1,
Kv2.1,
Shab
-289.0 Anti-sense Chr. 2 - 167095969 - 167188818 on NCBI Build GRCm38
Downstream prostaglandin I2 (prostacyclin) synthase Ptgis Cyp8,
Cyp8a1,
Pgis
10.7kb Anti-sense Chr. 2 - 167203194 - 167252127 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0