This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01247218_m1
See other INPP4B GE Assays ›
Gene Symbol:
Inpp4b
Gene Name:
inositol polyphosphate-4-phosphatase, type II
Gene Aliases:
E130107I17Rik
Chromosome Location:
Chr.8: 81342342 - 82152019 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01247218_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Inpp4b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001024617.3 NP_001019788.1 18 - 19 2327 105
NM_001297591.1 NP_001284520.1 18 - 19 2304 105
NM_001297593.1 NP_001284522.1 17 - 18 2185 105
NM_001297596.1 NP_001284525.1 18 - 19 2304 105
GenBank mRNA AK053554.1 - 11 - 12 1456 105
AK089068.1 - 13 - 14 1558 105
AK139397.1 - 11 - 12 1804 105
AK141502.1 - 20 - 21 2301 105
BC044876.1 - 4 - 5 691 105
BC064813.1 - 16 - 17 2029 6826283 105
DQ296611.1 - 18 - 19 2304 105
DQ296612.1 - 17 - 18 2185 105
DQ296613.1 - 18 - 19 2304 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800000899.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Inpp4b

Gene Ontology Categories:

Function(s) Process(es)

cellular calcium ion homeostasis
dephosphorylation
phosphatidylinositol-3-phosphate biosynthetic process
negative regulation of osteoclast differentiation
regulation of nucleocytoplasmic transport
regulation of bone remodeling
inositol phosphate dephosphorylation
phosphatidylinositol dephosphorylation
regulation of protein kinase B signaling
lipid binding
phosphatidylinositol-3,4-bisphosphate 4-phosphatase activity
hydrolase activity
phosphatidylinositol bisphosphate phosphatase activity
phosphatidylinositol trisphosphate phosphatase activity
phosphatidylinositol-4,5-bisphosphate 4-phosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800006704.mm.1 NM_001024617.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800006704.mm.1 TC0800000899.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800006703.mm.1 NM_001024617.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800006703.mm.1 TC0800000899.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0