This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01246420_m1
See other NECAB3 GE Assays ›
Gene Symbol:
Necab3
Gene Name:
N-terminal EF-hand calcium binding protein 3
Gene Aliases:
2900010M17Rik, AI853434, Apba2bp, Nip1, XB51
Chromosome Location:
Chr.2: 154544399 - 154559406 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01246420_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Necab3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021546.3 NP_067521.2 1 - 2 114 107
GenBank mRNA AK013520.1 - 1 - 2 113 107
AK044296.1 - 1 - 2 106 107
AK158225.1 - 1 - 2 116 107

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200004941.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Necab3

Gene Ontology Categories:

Function(s) Process(es)

protein metabolic process
regulation of amyloid precursor protein biosynthetic process
calcium ion binding
protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200041918.mm.1 NM_021546.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200041918.mm.1 NM_021546.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200041918.mm.1 TC0200004941.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200041920.mm.1 NM_021546.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200041920.mm.1 NM_021546.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200041920.mm.1 TC0200004941.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0