This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01245502_m1
See other SLC2A4 GE Assays ›
Gene Symbol:
Slc2a4
Gene Name:
solute carrier family 2 (facilitated glucose transporter), member 4
Gene Aliases:
Glut-4, Glut4
Chromosome Location:
Chr.11: 69942286 - 69948221 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01245502_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc2a4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009204.2 NP_033230.2 1 - 2 242 79
GenBank mRNA AB008453.1 - 1 - 2 40 79
AK137607.1 - 1 - 2 227 79
AK138832.1 - 1 - 2 216 79
AK160882.1 - 1 - 2 228 79
AK164310.1 - 1 - 2 217 79
BC014282.1 - 1 - 2 212 4207674 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003078.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc2a4

Gene Ontology Categories:

Function(s) Process(es)

transport
carbohydrate transport
amylopectin biosynthetic process
glucose transport
proton transport
cellular response to insulin stimulus
hexose transmembrane transport
glucose homeostasis
glucose import
brown fat cell differentiation
transmembrane transport
cellular response to hypoxia
cellular response to osmotic stress
cation transmembrane transport
transporter activity
glucose transmembrane transporter activity
insulin-responsive hydrogen:glucose symporter activity
protein binding
transmembrane transporter activity
substrate-specific transmembrane transporter activity
D-glucose transmembrane transporter activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100028330.mm.1 NM_009204.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028330.mm.1 NM_009204.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100028330.mm.1 TC1100003078.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100028334.mm.1 NM_009204.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028334.mm.1 NM_009204.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100028334.mm.1 TC1100003078.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100028332.mm.1 NM_009204.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028332.mm.1 NM_009204.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100028332.mm.1 TC1100003078.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100028333.mm.1 NM_009204.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028333.mm.1 NM_009204.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100028333.mm.1 TC1100003078.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 913136 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0