This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01245423_m1
See other ALS2CL GE Assays ›
Gene Symbol:
Als2cl
Gene Name:
ALS2 C-terminal like
Gene Aliases:
79H19D, 79H19E, BC022645, D930044G19Rik, mRn.49018
Chromosome Location:
Chr.9: 110880136 - 110900530 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01245423_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Als2cl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001146059.1 NP_001139531.1 8 - 9 979 68
NM_001146060.1 NP_001139532.1 8 - 9 1059 68
NM_146228.4 NP_666340.3 8 - 9 976 68
GenBank mRNA AB107016.1 - 8 - 9 976 68
AF481142.1 - 7 - 8 872 68
AK053076.1 - 8 - 9 949 68
AK131149.1 - 8 - 9 978 68
AK155451.1 - 8 - 9 1045 68
BC117928.1 - 7 - 8 929 40097635 68
BC117929.1 - 7 - 8 926 40097636 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001475.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Als2cl

Gene Ontology Categories:

Function(s) Process(es)

endosome organization
protein localization
regulation of Rho protein signal transduction
Rho guanyl-nucleotide exchange factor activity
GTPase activator activity
protein binding
Rab guanyl-nucleotide exchange factor activity
Rab GTPase binding
identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900012363.mm.1 NM_001146059.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900012363.mm.1 NM_001146059.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900012363.mm.1 NM_001146060.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900012363.mm.1 NM_001146060.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900012363.mm.1 NM_146228.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900012363.mm.1 NM_146228.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900012363.mm.1 TC0900001475.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header