This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01245409_m1
See other ALS2CL GE Assays ›
Gene Symbol:
Als2cl
Gene Name:
ALS2 C-terminal like
Gene Aliases:
79H19D, 79H19E, BC022645, D930044G19Rik, mRn.49018
Chromosome Location:
Chr.9: 110880136 - 110900530 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01245409_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Als2cl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001146059.1 NP_001139531.1 15 - 16 1742 123
NM_001146060.1 NP_001139532.1 15 - 16 1822 123
NM_146228.4 NP_666340.3 15 - 16 1739 123
GenBank mRNA AB107016.1 - 15 - 16 1739 123
AF481142.1 - 14 - 15 1635 123
AK030815.1 - 8 - 9 876 123
AK053076.1 - 15 - 16 1712 123
AK131149.1 - 15 - 16 1741 123
AK155451.1 - 15 - 16 1808 123
BC117928.1 - 14 - 15 1692 40097635 123
BC117929.1 - 14 - 15 1689 40097636 123

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK030815.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Als2cl

Gene Ontology Categories:

Function(s) Process(es)

endosome organization
protein localization
regulation of Rho protein signal transduction
Rho guanyl-nucleotide exchange factor activity
GTPase activator activity
protein binding
Rab guanyl-nucleotide exchange factor activity
Rab GTPase binding
identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900012371.mm.1 NM_001146059.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900012371.mm.1 NM_001146059.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900012371.mm.1 NM_001146060.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900012371.mm.1 NM_001146060.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900012371.mm.1 NM_146228.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900012371.mm.1 NM_146228.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900012371.mm.1 TC0900001475.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900012372.mm.1 NM_001146059.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900012372.mm.1 NM_001146059.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900012372.mm.1 NM_001146060.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900012372.mm.1 NM_001146060.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900012372.mm.1 NM_146228.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900012372.mm.1 NM_146228.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900012372.mm.1 TC0900001475.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0