Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800008915.mm.1 |
NM_133954.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008915.mm.1 |
NM_133954.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008915.mm.1 |
TC0800001132.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800008919.mm.1 |
NM_133954.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008919.mm.1 |
NM_133954.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008919.mm.1 |
TC0800001132.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800008917.mm.1 |
NM_133954.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008917.mm.1 |
NM_133954.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008917.mm.1 |
TC0800001132.mm.1 |
MTA TC |