This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01243518_m1
See other DDX19B GE Assays ›
Gene Symbol:
Ddx19b
Gene Name:
DEAD (Asp-Glu-Ala-Asp) box polypeptide 19b
Gene Aliases:
2810457M08Rik, 4921519L13Rik, AW260119
Chromosome Location:
Chr.8: 111003186 - 111031751 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01243518_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ddx19b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001190786.1 NP_001177715.1 2 - 3 182 81
NM_001190800.1 NP_001177729.1 2 - 3 225 81
NM_172284.5 NP_758488.1 2 - 3 225 81
GenBank mRNA AK013362.1 - 2 - 3 220 81
AK033235.1 - 2 - 3 183 81
AK147499.1 - 2 - 3 220 81
BC025594.1 - 2 - 3 185 5342859 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800002935.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ddx19b

Gene Ontology Categories:

Function(s) Process(es)

mRNA export from nucleus
biological_process
RNA secondary structure unwinding
molecular_function
ATP-dependent RNA helicase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800022594.mm.1 NM_001190786.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022594.mm.1 NM_001190786.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022594.mm.1 NM_001190800.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022594.mm.1 NM_001190800.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022594.mm.1 NM_172284.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022594.mm.1 NM_172284.5 Refseq
Affymetrix Mouse Clariom S Array PSR0800022594.mm.1 TC0800002935.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800022595.mm.1 NM_001190786.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022595.mm.1 NM_001190786.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022595.mm.1 NM_001190800.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022595.mm.1 NM_001190800.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022595.mm.1 NM_172284.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022595.mm.1 NM_172284.5 Refseq
Affymetrix Mouse Clariom S Array PSR0800022595.mm.1 TC0800002935.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800022601.mm.1 NM_001190786.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022601.mm.1 NM_001190786.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022601.mm.1 TC0800002935.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800022596.mm.1 NM_001190786.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022596.mm.1 NM_001190786.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022596.mm.1 NM_001190800.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022596.mm.1 NM_001190800.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022596.mm.1 NM_172284.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022596.mm.1 NM_172284.5 Refseq
Affymetrix Mouse Clariom S Array PSR0800022596.mm.1 TC0800002935.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0