This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01241479_m1
See other RNF157 GE Assays ›
Gene Symbol:
Rnf157
Gene Name:
ring finger protein 157
Gene Aliases:
2610036E23Rik, A130073L17Rik, mKIAA1917
Chromosome Location:
Chr.11: 116336345 - 116413032 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01241479_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rnf157
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027258.1 NP_081534.1 13 - 14 1665 92
GenBank mRNA AK038042.1 - 7 - 8 892 92
AK122571.1 - 3 - 4 401 92
AK169566.1 - 8 - 9 845 92
BC053070.1 - 13 - 14 1444 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100004115.mm.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
zinc ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100038932.mm.1 NM_027258.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100038932.mm.1 NM_027258.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100038932.mm.1 TC1100004115.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100038933.mm.1 NM_027258.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100038933.mm.1 NM_027258.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100038933.mm.1 TC1100004115.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0