This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01241314_m1
See other FLOT2 GE Assays ›
Gene Symbol:
Flot2
Gene Name:
flotillin 2
Gene Aliases:
AI573412, Esa, reggie-2
Chromosome Location:
Chr.11: 78037868 - 78060434 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01241314_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Flot2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001040403.1 NP_001035493.1 4 - 5 462 108
NM_001284227.1 NP_001271156.1 5 - 6 546 108
NM_001284228.1 NP_001271157.1 5 - 6 409 108
NM_008028.2 NP_032054.1 5 - 6 553 108
GenBank mRNA AK004589.1 - 4 - 5 400 108
AK030199.1 - 4 - 5 425 108
AK149530.1 - 5 - 6 484 108
AK170557.1 - 4 - 5 463 108
AK187814.1 - 4 - 5 346 108
AK213895.1 - 4 - 5 317 108
BC070423.1 - 5 - 6 511 6403816 108
U07890.1 - 5 - 6 423 108

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001140.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Flot2

Gene Ontology Categories:

Function(s) Process(es)

membrane raft assembly
cell adhesion
negative regulation of gene expression
protein localization to plasma membrane raft
protein stabilization
establishment of protein localization to plasma membrane
negative regulation of amyloid precursor protein catabolic process
positive regulation of establishment of T cell polarity
protease binding
ionotropic glutamate receptor binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100010479.mm.1 NM_001040403.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100010479.mm.1 NM_001040403.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100010479.mm.1 NM_001284227.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100010479.mm.1 NM_001284228.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100010479.mm.1 NM_008028.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100010479.mm.1 NM_008028.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100010479.mm.1 TC1100001140.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0