This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01239632_m1
See other CPSF3 GE Assays ›
Gene Symbol:
Cpsf3
Gene Name:
cleavage and polyadenylation specificity factor 3
Gene Aliases:
-
Chromosome Location:
Chr.12: 21286297 - 21315056 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01239632_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cpsf3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018813.3 NP_061283.2 15 - 16 1984 89
GenBank mRNA AK004317.1 - 2 - 3 194 89
AK010566.1 - 8 - 9 1051 89
AK045238.1 - 4 - 5 354 89
AK149953.1 - 15 - 16 1911 89
AK157205.1 - 15 - 16 1926 89
AK158036.1 - 15 - 16 1897 89
AK170842.1 - 15 - 16 1869 89
AK171838.1 - 15 - 16 1915 89
BC006748.1 - 10 - 11 1539 89
BC023297.1 - 15 - 16 1870 4192224 89

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cpsf3

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
mRNA 3'-end processing by stem-loop binding and cleavage
RNA phosphodiester bond hydrolysis, endonucleolytic
RNA binding
nuclease activity
endonuclease activity
endoribonuclease activity
5'-3' exonuclease activity
hydrolase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0