This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01238923_g1
See other BAMBI GE Assays ›
Gene Symbol:
Bambi
Gene Name:
BMP and activin membrane-bound inhibitor
Gene Aliases:
2610003H06Rik
Chromosome Location:
Chr.18: 3507957 - 3516404 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01238923_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bambi
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026505.2 NP_080781.1 2 - 3 619 62
GenBank mRNA AK011305.1 - 2 - 3 642 62
AK045192.1 - 2 - 3 619 62
AK143185.1 - 2 - 3 634 62
AK164632.1 - 2 - 3 622 62
BC019378.1 - 2 - 3 601 5009872 62

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bambi

Gene Ontology Categories:

Function(s) Process(es)

transforming growth factor beta receptor signaling pathway
positive regulation of cell proliferation
regulation of cell shape
positive regulation of epithelial to mesenchymal transition
cell migration
negative regulation of transforming growth factor beta receptor signaling pathway
positive regulation of protein binding
positive regulation of catenin import into nucleus
positive regulation of transcription, DNA-templated
positive regulation of canonical Wnt signaling pathway
frizzled binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800000064.mm.1 NM_026505.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800000064.mm.1 NM_026505.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800000064.mm.1 TC1800000008.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800000065.mm.1 NM_026505.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800000065.mm.1 NM_026505.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800000065.mm.1 TC1800000008.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0