This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01236498_m1
See other IGHMBP2 GE Assays ›
Gene Symbol:
Ighmbp2
Gene Name:
immunoglobulin mu binding protein 2
Gene Aliases:
AEP, Catf1, RIPE3b1, Smbp-2, Smbp2, Smubp2, nmd, sma
Chromosome Location:
Chr.19: 3259076 - 3283010 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01236498_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ighmbp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009212.2 NP_033238.2 11 - 12 1855 57
GenBank mRNA AK142456.1 - 11 - 12 1659 57
AK153763.1 - 11 - 12 1816 57
AK154805.1 - 11 - 12 1852 57
AK155899.1 - 11 - 12 1852 57
BC052025.1 - 10 - 11 1727 57
BC060085.1 - 10 - 11 1727 57
BC066215.1 - 11 - 12 1798 57
L10075.1 - 11 - 12 1751 57

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000878.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ighmbp2

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
translation
spinal cord motor neuron differentiation
DNA duplex unwinding
protein homooligomerization
tRNA binding
nucleotide binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
nucleic acid binding
DNA binding
RNA binding
helicase activity
ATP binding
DNA-dependent ATPase activity
transcription factor binding
RNA-dependent ATPase activity
zinc ion binding
hydrolase activity
ATP-dependent 5'-3' RNA helicase activity
ribosome binding
ATP-dependent 5'-3' DNA helicase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900007533.mm.1 NM_009212.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900007533.mm.1 NM_009212.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900007533.mm.1 TC1900000878.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0