This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01236497_m1
See other NDUFS8 GE Assays ›
Gene Symbol:
Ndufs8
Gene Name:
NADH dehydrogenase (ubiquinone) Fe-S protein 8
Gene Aliases:
BC021616, CI-23kD, TYKY
Chromosome Location:
Chr.19: 3908863 - 3912774 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01236497_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndufs8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271443.1 NP_001258372.1 1 - 2 67 66
GenBank mRNA AK135069.1 - 1 - 2 68 66
AK153900.1 - 1 - 2 44 66
AK164901.1 - 1 - 2 44 66
AK167907.1 - 1 - 2 45 66

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK135069.1, AK153900.1, AK164901.1, AK167907.1, NM_001271443.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

transport
response to oxidative stress
mitochondrial respiratory chain complex I assembly
oxidation-reduction process
NADH dehydrogenase activity
NADH dehydrogenase (ubiquinone) activity
oxidoreductase activity
oxidoreductase activity, acting on NAD(P)H
metal ion binding
iron-sulfur cluster binding
4 iron, 4 sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0