This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01236212_g1
See other STIP1 GE Assays ›
Gene Symbol:
Stip1
Gene Name:
stress-induced phosphoprotein 1
Gene Aliases:
Hop, Sti1, p60
Chromosome Location:
Chr.19: 7020696 - 7040026 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01236212_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stip1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016737.2 NP_058017.1 3 - 4 499 117
GenBank mRNA AK075988.1 - 3 - 4 437 117
AK088494.1 - 3 - 4 441 117
AK134171.1 - 3 - 4 415 117
AK149493.1 - 3 - 4 415 117
AK161645.1 - 3 - 4 418 117
AK167273.1 - 3 - 4 433 117
AK168181.1 - 3 - 4 415 117
U27830.1 - 3 - 4 416 117

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900001031.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stip1

Gene Ontology Categories:

Process(es)

protein binding
protein C-terminus binding
Hsp70 protein binding
poly(A) RNA binding
chaperone binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900009631.mm.1 NM_016737.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009631.mm.1 NM_016737.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900009631.mm.1 TC1900001031.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900009632.mm.1 NM_016737.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009632.mm.1 NM_016737.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900009632.mm.1 TC1900001031.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900009629.mm.1 NM_016737.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900009629.mm.1 NM_016737.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900009629.mm.1 TC1900001031.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0