This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01235633_m1
See other GATA6 GE Assays ›
Gene Symbol:
Gata6
Gene Name:
GATA binding protein 6
Gene Aliases:
AA410133, GATA-6
Chromosome Location:
Chr.18: 11052475 - 11085636 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01235633_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gata6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010258.3 NP_034388.2 1 - 2 233 58
GenBank mRNA AK053151.1 - 1 - 2 236 58
AK142381.1 - 1 - 2 93 58
BY084328.1 - 1 - 2 273 58

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK142381.1, NM_010258.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gata6

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
in utero embryonic development
liver development
outflow tract septum morphogenesis
type B pancreatic cell differentiation
pancreatic A cell differentiation
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
phospholipid metabolic process
endodermal cell fate determination
regulation of gene expression
cardiac muscle hypertrophy in response to stress
positive regulation of BMP signaling pathway
epithelial cell differentiation
pancreas development
negative regulation of transforming growth factor beta1 production
negative regulation of transforming growth factor beta2 production
tube morphogenesis
endodermal cell differentiation
response to drug
positive regulation of apoptotic process
negative regulation of apoptotic process
response to estrogen
positive regulation of angiogenesis
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
organ formation
cardiac muscle tissue development
smooth muscle cell differentiation
cardiac muscle cell differentiation
positive regulation of cardiac muscle cell proliferation
lung saccule development
Clara cell differentiation
Type II pneumocyte differentiation
intestinal epithelial cell differentiation
cardiac vascular smooth muscle cell differentiation
response to growth factor
positive regulation of cell cycle arrest
cellular response to gonadotropin stimulus
cellular response to hypoxia
cellular response to BMP stimulus
RNA polymerase II core promoter sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
RNA polymerase II repressing transcription factor binding
enhancer sequence-specific DNA binding
DNA binding
chromatin binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
transcription factor binding
zinc ion binding
protein kinase binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800000593.mm.1 NM_010258.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800000593.mm.1 NM_010258.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800000593.mm.1 TC1800000091.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800000594.mm.1 NM_010258.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800000594.mm.1 NM_010258.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800000594.mm.1 TC1800000091.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 500605 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0