This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01233792_m1
See other CNOT2 GE Assays ›
Gene Symbol:
Cnot2
Gene Name:
CCR4-NOT transcription complex, subunit 2
Gene Aliases:
2600016M12Rik, 2810470K03Rik, AA537049, AA959607, AW557563, C79650
Chromosome Location:
Chr.10: 116485161 - 116581535 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01233792_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cnot2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037846.3 NP_001032935.2 7 - 8 804 106
NM_001037847.2 NP_001032936.2 7 - 8 734 106
NM_028082.2 NP_082358.2 6 - 7 681 106
NR_024327.1 5 - 6 614 106
GenBank mRNA AK011231.1 - 5 - 6 604 106
AK078121.1 - 7 - 8 794 106
AK149767.1 - 7 - 8 778 106
AK167834.1 - 7 - 8 782 106
BC043133.1 - 7 - 8 705 106
BC063105.1 - 8 - 9 947 5003026 106
BC065171.1 - 7 - 8 734 6415034 106
BC090624.1 - 5 - 6 610 6815664 106

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002907.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
trophectodermal cell differentiation
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of translation
multicellular organism development
positive regulation of cytoplasmic mRNA processing body assembly
gene silencing by RNA
negative regulation of intracellular estrogen receptor signaling pathway
RNA phosphodiester bond hydrolysis, exonucleolytic
regulation of stem cell population maintenance
RNA polymerase II transcription corepressor binding
poly(A)-specific ribonuclease activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000021975.mm.1 NM_001037846.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000021975.mm.1 NM_001037846.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000021975.mm.1 NM_001037847.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000021975.mm.1 NM_001037847.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000021975.mm.1 NM_028082.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000021975.mm.1 NM_028082.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000021975.mm.1 NR_024327.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000021975.mm.1 NR_024327.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000021975.mm.1 TC1000002907.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000021976.mm.1 NM_001037846.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000021976.mm.1 NM_001037846.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000021976.mm.1 NM_001037847.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000021976.mm.1 NM_001037847.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000021976.mm.1 NM_028082.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000021976.mm.1 NM_028082.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000021976.mm.1 NR_024327.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000021976.mm.1 NR_024327.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000021976.mm.1 TC1000002907.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-77574795ff-5hfml:80/100.66.128.142:80.
git-commit: 5cc8a939e1477954742eeb281a87e54f28adfb77
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header