Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000021978.mm.1 |
NM_001037846.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000021978.mm.1 |
NM_001037846.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000021978.mm.1 |
NM_001037847.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000021978.mm.1 |
NM_001037847.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000021978.mm.1 |
NM_028082.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000021978.mm.1 |
NM_028082.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000021978.mm.1 |
NR_024327.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000021978.mm.1 |
NR_024327.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000021978.mm.1 |
TC1000002907.mm.1 |
MTA TC |