Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700022384.mm.1 |
NM_013933.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022384.mm.1 |
NM_013933.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022384.mm.1 |
TC1700002411.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700022383.mm.1 |
NM_013933.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022383.mm.1 |
NM_013933.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022383.mm.1 |
TC1700002411.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700022385.mm.1 |
NM_013933.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022385.mm.1 |
NM_013933.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022385.mm.1 |
TC1700002411.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700022386.mm.1 |
NM_013933.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022386.mm.1 |
NM_013933.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022386.mm.1 |
TC1700002411.mm.1 |
MTA TC |