This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01229131_m1
See other CEBPZ GE Assays ›
Gene Symbol:
Cebpz
Gene Name:
CCAAT/enhancer binding protein zeta
Gene Aliases:
AI848081, CBF2, Cbf, Cebpa-rs1
Chromosome Location:
Chr.17: 78919003 - 78937070 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01229131_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cebpz
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001024806.2 NP_001019977.1 5 - 6 2200 123
GenBank mRNA AK145374.1 - 5 - 6 2182 123
AK155990.1 - 5 - 6 2183 123
AK158771.1 - 5 - 6 2177 123
AK159081.1 - 5 - 6 2197 123
U19891.1 - 5 - 6 2171 123
U19892.1 - 5 - 6 1885 123

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700002561.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cebpz

Panther Classification:

Molecular Function -

DNA-binding transcription factor gene-specific transcriptional regulator

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700023443.mm.1 NM_001024806.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700023443.mm.1 NM_001024806.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700023443.mm.1 TC1700002561.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0