This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01227083_m1
See other RMDN2 GE Assays ›
Gene Symbol:
Rmdn2
Gene Name:
regulator of microtubule dynamics 2
Gene Aliases:
AW061290, Fam82a1, RMD-2, mRMD-2
Chromosome Location:
Chr.17: 79578598 - 79685067 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01227083_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rmdn2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_201361.2 NP_958749.1 5 - 6 899 73
GenBank mRNA AK034617.1 - 5 - 6 901 73
AK139382.1 - 5 - 6 1118 73
BC024059.1 - 5 - 6 979 5067179 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001197.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rmdn2

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700011301.mm.1 TC1700001197.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700011306.mm.1 NM_201361.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700011306.mm.1 NM_201361.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700011306.mm.1 TC1700001197.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0